Stampa…

n-Esano, AR®, Laboratory Reagent, Macron Fine Chemicals™

Fornitore: Avantor Performance Materials

Synonyms: Hexyl hydride, Hexanen, Hexane

6729-25EA 161.5 EUR
6729-25 6729-45
n-Esano, AR®, Laboratory Reagent, Macron Fine Chemicals™
n-Esano
Formula: H₃C(CH₂)₄CH₃
Peso molecolare: 86.18 g/mol
Punto di ebollizione: 69
Punto di fusione: -94.3
Flash Pt: -22
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR