Stampa…
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide CN Fused-Core®
Fornitore: Merck
66965-U 66966-U 66967-U 66968-U 66969-U 66970-U 66971-U 66972-U 66973-U 66974-U 66975-U 66976-U 66977-U 66978-U 66979-U 67061-U 67062-U 67063-U 67064-U 67065-U 67066-U 67067-U 67068-U 67069-U 67071-U 67080-U 67081-U 67082-U 67083-U 67084-U 67159-U 67160-U 67161-U 67163-U 67164-U 67165-U
66965-UEA 596 EUR
66965-U 66966-U 66967-U 66968-U 66969-U 66970-U 66971-U 66972-U 66973-U 66974-U 66975-U 66976-U 66977-U 66978-U 66979-U 67061-U 67062-U 67063-U 67064-U 67065-U 67066-U 67067-U 67068-U 67069-U 67071-U 67080-U 67081-U 67082-U 67083-U 67084-U 67159-U 67160-U 67161-U 67163-U 67164-U 67165-U
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide CN Fused-Core®
Colonne cromatografiche
BIOshell™ 160 Å Peptide CN columns provide an alternative selectivity to the traditional C18 bonded phases.
  • Higher resolutions and higher peak capacities of peptides at ~50% backpressures of sub-2 µm fully porous paticles (FPP)-packed columns
  • Lower backpressure allows for columns to be used in series to maximise resolutions and peak capacity of complex proteomic samples or tryptic digests
  • Compatible with all UHPLC and LC-MS instrumentation
The introduction of the cyano group enables peptides and small proteins to be separatedd due to a combination ofMore Product Information