Stampa…
Ion selective electrodes, chlorine, Orion™
Fornitore: Thermo Orion
9770BNWP
662-1595EA 674 EUR
662-1595
Ion selective electrodes, chlorine, Orion™
Elettrodi Elettrodi ISE
These electrodes are designed for measuring total residual chlorine quickly and accurately.
  • All forms of total residual chlorine are measured
  • Combination electrode does not require a separate reference electrode, making it convenient to use
Informazioni per l'ordine: The electrode with screw cap is supplied without cable.