Stampa…

Metanolo ≥99.5% NF, Ph. Eur., BP

Fornitore: VWR Chemicals
Pericolo

Synonyms: Alcol metilico, Alcol denaturato

6520.9025EA 223 EUR
6520.9025 6520.9200
Metanolo ≥99.5% NF, Ph. Eur., BP
Metanolo

Appearance: Clear, colourless, volatile, hygroscopic liquid.
Residues of metal catalysts or reagents: Conforms to EMEA/CHMP/SWP/4446/2000
(*) Absorption curve: The spectrum shows a steadily descending curve with no observable peaks or shoulders.


Attenzione: Not for use as Active Pharmaceutical Ingredient (API)

Formula: CH₃OH
Punto di ebollizione: 64,6 °C (1013 hPa)
Punto di fusione: –98 °C
Densità: 0,7918 g/cm³ (20 °C)
Flash Pt: 11 °C (coppa chiusa)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Conforms (see CoA/CoS)
Refractive index (20°C) 1.328 --> 1.330
Residual solvents (CPMP/ICH/283/95) Conforms
Residues of metal catalysts or reagents Unlikely by manuf.process
As (Arsenic) Max. 2 ppm
Cd (Cadmium) Max. 0.1 ppm
Co (Cobalt) Max. 1 ppm
Hg (Mercury) Max. 2 ppm
Ni (Nickel) Max. 6 ppm
Pb (Lead) Max. 2 ppm
V (Vanadium) Max. 10 ppm
Conforms to Ph.Eur./NF/BP Passes test
Ph.Eur./BP specifications: Max. 0.02
Identification A Passes test NF
Identification B Passes test NF
Appearance test Passes test Ph.Eur./BP
Acidity or alkalinity Passes test Ph.Eur./BP
Relative density [Ph.Eur./BP] 0.791 --> 0.793
Absorbance (230 nm) [Ph.Eur./BP] Max. 0.15
Absorbance (250 nm) [Ph.Eur./BP] Max. 0.05
Absorbance (270 nm) [Ph.Eur/BP] Max. 0.02
Absorbance (290 nm) [Ph.Eur./BP] Max. 0.01
Absorption curve (*) Passes test Ph.Eur./BP
Impurity A (Benzene V/V) [Ph.Eur./BP] Max. 2 ppm
Any impurity [Ph.Eur./BP] Max. 0.1 %
Total impurities [Ph.Eur./BP] Max. 0.3 %
Disregard limit [Ph.Eur./BP] Max. 50 ppm
Reducing substances Passes test Ph.Eur./BP
Residue on evaporation [Ph.Eur./BP] Max. 10 ppm
Water [Ph.Eur./BP] Max. 0.10 %
NF specifications:
Assay [NF] Min. 99.5 %
Non-volatile residue [NF] Max. 0.001 %
Acetone and aldehydes (as acetone) [NF] Max. 0.003 %
Readily carbonisable substances Passes test NF
Readily oxidisable substances Passes test NF
Acidity Passes test NF
Alkalinity (as ammonia) [NF] Max. 3 ppm
Water determination [NF] Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR