Stampa…
Microviscosimetro Ubbelohde
Fornitore: SI Analytics
285401017 285401222 285401025 285401033 285401128 285401136 285401214 285401239 285401247 285401009 285401206 285401041
645-0071EA 561 EUR
645-0071 645-0077 645-0072 645-0073 616-3358 616-3359 645-0076 645-0078 645-0079 645-0070 645-0075 645-0074
Microviscosimetro Ubbelohde
Viscosimetri Viscometri Capillari
Viscometers, with suspended ball level, for determination of relative or absolute kinematic viscosity.
  • Suitable for small samples and short running times
  • Ring marks for checking by manually
  • Uncalibrated or calibrated with constant for automatic measurements
Certificazioni: DIN 51 562-2