Stampa…
Staining jar acc.to Coplin
Fornitore: DELTALAB
191087
631-9730EA 36.6 EUR
631-9730
Staining jar acc.to Coplin
Contenitori Vaschette di colorazione
Jar made of PP. Rectangular container with rounded base and screw ribbed flat cap with an inner ring for preventing leakage.
  • Unbreakable
  • Ideal for used as a storage system
  • Good chemical and temperature resistance
These staining jar hold up to 10 slides (Two per groove).