Stampa…
Optical Glass Filters
Fornitore: SCHOTT AG Lighting and imaging
258306 158211 258302 258303 258304 258305 258307 158207 258313 258330
SCOC258306EA 48 EUR
SCOC258306 631-0010 631-0523 631-0524 631-0525 631-0526 631-0916 630-1459 SCOC258313 SCOC258330
Optical Glass Filters
Filtri Filtri ottici
Optical glass filters are used to change the light color of the emitted light from a KL Cold lightsource.
  • Insert Filters for cold light source with filter holder
  • Glass Optical Filters for better temperature resistance
  • Color filters with high transmission and unaffected by light entrance angle
  • The Daylight Filter for Halogen Light Sources Increases the color temperature from a warm halogen light to a cold daylight
  • The Halogen Filter for LED lightsources decrease the color temperature from a cold daylight to a warm halogen light
  • The (empty) Filter Holder for