Stampa…
Folding magnifier, metal
Fornitore: ESCHENBACH OPTIK
11766
630-8231EA 75.2 EUR
630-8231
Folding magnifier, metal
Lenti d'ingrandimento
Metal folding magnifier with aplanatic lens and 6× magnification.
  • Lens made of high quality silicate
  • Matt chromium-plated brass body with eyelet
  • Black painted metal lens frame
  • Distortion-free illustration