Stampa…
LED cold light source, KL 300 LED (without fan)
Fornitore: SCHOTT AG Lighting and imaging
120300
630-1528EA 445 EUR
630-1528
LED cold light source, KL 300 LED (without fan)
Luci Illuminatori a luce fredda
This compact LED cold light source for eco-friendly PURAVIS® light guides is driven by a single LED and provides a brightness equivalent to a 30 W halogen lamp and is therefore ideal as a standard illumination system for the requirements of routine inspections and education.
  • Ecological lead-free fibre optics light guides for more visible and purer images
  • 80 lm lightflux
  • Long lifetime of at least 50000 hours
  • No fan, silent operation
  • Compact ergonomical design
  • Mountable direct to any microscope stand or pillar
  • Wide range power supply with international clip