Stampa…
Lenti di ingrandimento da lettura, easyPOCKET
Fornitore: ESCHENBACH OPTIK
152110 152122
630-1037EA 88.5 EUR
630-1037 630-1321
Lenti di ingrandimento da lettura, easyPOCKET
Lenti d'ingrandimento
These pocket magnifiers provide magnification and illumination in a credit card format.
  • Ultra-flat and lightweight design
  • Diffractive aspheric PXM® lightweight lens
  • SMD LED illumination, which is switched on when you slide the lens out of its case
  • Easy handling - No bulb change required
Informazioni per l'ordine: Supplied with two button cells.
Informazioni per l'ordine: Supplied with two button cells.