Stampa…

Litio carbonato ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS per microscopia, Fluka™

Fornitore: Honeywell Chemicals
Attenzione

62470-100G 62470-500G
62470-100GEA 51.39 EUR
62470-100G 62470-500G
Litio carbonato ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS per microscopia, Fluka™
Litio carbonato
Formula: Li₂CO₃
Punto di ebollizione: 1,300 °C
Punto di fusione: 720 °C
Densità: 2,110 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Visualizza i codici

Risultati del test di specificazione

Assay 99.0 - 101.0 %
Insoluble in HCl Max. 0.01 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 10 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (ICP) ACS Max. 0.002 %
S-compounds (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 20 ppm
Ammonium (NH₄) Max. 5 ppm
Nitrate (NO₃) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR