Stampa…
Multipurpose Centrifuge for Dairy Laboratory, SuperVario-N
Fornitore: GERBER FUNKE
3680
614-0174EA 5870 EUR
614-0174
Multipurpose Centrifuge for Dairy Laboratory, SuperVario-N
Centrifughe Centrifughe da banco
Bench top centrifuge with high versatility, offering four program settings: Dr. N. Gerber's method, Babcock's method (determination of fat content), Roese-Gottlieb's (determination of fat content, reference method), and Solubility (determination of powdered milk solubility).
  • Stainless steel housing
  • Programmable rotor speed from 600 to 1130 min⁻¹ (77 to 372 ×g) in increments of 10 min⁻¹
  • Programmable heater up to +68 °C in increments of 1 °C
  • Programmable centrifugation time from 1 to 99 minutes
  • Automatic lid locking, imbalance switch off, automatic brake