Stampa…
Laboratory syringes, pistol grip, Dosys™ 170 series
Fornitore: Socorex Isba
174.0501 174.0502 174.0505 174.0510 173.0501 173.0510 173.0520 173.0505 173.05003 173.05005 173.0502 172.0510 172.0505 172.0501 172.0502
613-4803EA 210 EUR
613-4803 613-4804 613-4805 613-4806 613-4809 613-4812 613-4813 613-4811 613-4807 613-4808 613-0398 SOCO172.0510 SOCO172.0505 SOCO172.0501 SOCO172.0502
Laboratory syringes, pistol grip, Dosys™ 170 series
Siringhe Siringhe, uso medico
Dosys™ 174 premium: Self-refilling, automatic syringe with detachable, Luer lock valve system. Enhanced corrosion resistance.
Dosys™ 173 classic: Self-refilling, automatic syringe with integrated, Luer lock valve system. Ideal for aquous, non corrosive liquids.
Dosys™ 172 basic: Automatic syringe without valve system. Luer lock nozzle.
Informazioni per l'ordine: Dosys™ 174 premium package includes syringe and 1 meter silicone feed tube, sinker, PVC protection sleeve, aspiration, vent and dosing cannulas, set of spare for valve, operating instructions. Dosys™ 173 classic packageMore Product Information