Stampa…
VWR®, Serological Pipettes, Open End
Fornitore: VWR Collection
612-5855EA 95.8 EUR
612-5855 612-5856 612-5857 612-5858 612-5859 612-5860 612-5861 612-5862
VWR®, Serological Pipettes, Open End
Pipette Pipette sierologiche
PS, transparent, graduated, non sterile or sterile. These disposable pipettes for liquid transfers are made of GPPS (general polystyrene), in compliance with USP Class VI.
  • DNase-/RNase-free, non pyrogenic, non cytotoxic, non haemolytic
  • Meet the requirement of BST/TSE; latex-free
  • Graduations are calibrated for accurate dispensing to within ±2%
  • Pipettes are colour coded according to their volume
  • Sterilised by irradiation
  • Every product is printed with a batch number for traceability
Manufactured in a Class 100 000 cleanroom environment. ManufacturedMore Product Information