Stampa…
Sterilisation Boxes for Pipettes
Fornitore: BOCHEM
8870 8871 8872 8873 8874
612-1626EA 213 EUR
612-1626 612-1627 612-1628 612-1629 612-1630
Sterilisation Boxes for Pipettes
Scatole Contenitori per sterilizzazione pipette
Stainless steel, with square base, autoclavable.
  • Can be autoclaved at 121 °C and sterilised at 205 °C
Bottom and underside of the lid are coated in silicone to minimise the risk of damaging the pipettes during transport and sterilisation