Stampa…

Acido cloridrico 36 - 38%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Fornitore: Avantor
Pericolo

Synonyms: Hydrochloric acid solution, Muriatic acid

6081.2500EA 62.92 EUR
6081.2500 6081.1000 6081.9025 6081.2500PE
Acido cloridrico 36 - 38%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Acido cloridrico
Formula: HCl
Punto di ebollizione: 110 °C (1013 hPa)
Punto di fusione: –30 °C
Densità: 1,18 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Assay 37 - 38 %
Ammonium (NH4) max. 3 ppm
Mercury (Hg) max. 5 ppb
Free Chlorine max. 0.5 ppm
Residue after Ignition max. 5 ppm
Sulfate (SO4) max. 1 ppm
Sulfite (SO3) max. 2 ppm
Trace Impurities (in ppm)
Silver (Ag) max. 0.01
Aluminium (Al) max. 0.05
Arsenic (As) max. 0.05
Barium (Ba) max. 0.02
Beryllium (Be) max. 0.01
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.02
Cobalt (Co) max. 0.01
Chromium (Cr) max. 0.05
Copper (Cu) max. 0.01
Iron (Fe) max. 0.20
Germanium (Ge) max. 0.1
Potassium (K) max. 0.10
Lithium (Li) max. 0.01
Magnesium (Mg) max. 0.05
Manganese (Mn) max. 0.01
Molybdenum (Mo) max. 0.02
Sodium (Na) max. 0.30
Nickel (Ni) max. 0.05
Lead (Pb) max. 0.05
Strontium (Sr) max. 0.01
Titanium (Ti) max. 0.05
Thallium (Tl) max. 0.05
Vanadium (V) max. 0.01
Zinc (Zn) max. 0.5
Zirconium (Zr) max. 0.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR