Stampa…

Magnesio cloruro esaidrato 99.0-102.0%, cristalli, AR® ACS, Macron Fine Chemicals™

Fornitore: Avantor Performance Materials

Synonyms: Magnesium dichloride

5958-04EA 119 EUR
5958-04 5958-26
Magnesio cloruro esaidrato 99.0-102.0%, cristalli, AR® ACS, Macron Fine Chemicals™
Magnesio cloruro esaidrato

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: MgCl₂·6H₂O
Peso molecolare: 203.3 g/mol
Punto di ebollizione: 1412
Punto di fusione: 117
Densità: 1.57 g/cm³ (25 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Visualizza i codici

Risultati del test di specificazione

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.0 - 102.0%
ACS - Assay (MgCl₂·6H₂O) 99.0 - 102.0 %
ACS - Ammonium (NH₄) ≤0.001 %
ACS - Barium (Ba) ≤0.005 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Manganese (Mn) ≤5.000 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - pH of 5% Solution 4.5 - 7.0
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Potassium (K) ≤0.005 %
ACS - Sodium (Na) ≤0.005 %
ACS - Strontium (Sr) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.002 %
The net weight of the product in each container is within +/- 2% of the netweight stated on the label.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR