Stampa…
Tubing
Fornitore: S.C.A.T.
108015 108016 108017 108018 108019 300021 300022 461053 461054 461055 461056
590-0705EA 105 EUR
590-0705 590-0706 590-0707 590-0708 590-0709 590-0827 590-0828 590-0829 590-0830 590-0831 SCTV461056
Tubing
Tubazioni Tubi per cromatografia
Flexible tubing made of PP and PTFE-EL material.
  • Available in various materials, lengths and properties
Electrically conductive plastic tubing provides safe protection against ignition hazards and electrostatic charging.