Stampa…
Distributor for flushing bottles, b.safe
Fornitore: Bohlender
M181-03 M181-10
590-0095EA 232 EUR
590-0095 590-0096
Distributor for flushing bottles, b.safe
Raccordi e connettori
Screw cap made of PPS for GL 45 thread / PP for GLS 80 thread, free movable insert made of PP with 4 lateral connectors (GL 45) suitable for b.safe Caps GL 45 and 3 centric capillary connectors (UNF ¹/₄"). Version and scope of delivery as per below chart.
  • No twisting of capillaries through 360° rotation of PP inserts
  • Opportunity to mark and stick
  • Inclined connectors for easy insertion of capillaries and suction filter
  • Integrated flat sealing and sealing lip for tight closure
Flushing the HPLC installation without pollution or loss of solvents. TheMore Product Information