Stampa…
Supelco® Ascentis® C8 HPLC Columns
Fornitore: Merck
581400-U 581401-U 581402-U 581407-U 581408-U 581414-U 581424-U 581425-U 581426-U 581427-U 581442-U
581400-UEA 584 EUR
581400-U 581401-U 581402-U 581407-U 581408-U 581414-U 581424-U 581425-U 581426-U 581427-U 581442-U
Supelco® Ascentis® C8 HPLC Columns
Colonne cromatografiche
Ascentis® C8 HPLC columns are optimised to the three terms of the resolution equation: efficiency, retention, and selectivity.
  • High purity, type B silica for inertness, reproducibility, and stability
  • Modern bonding process optimises bonded phase coverage and maximises stability, while minimising bleed and unwanted secondary interactions
  • Compatible with LC-MS and all of today's sensitive instruments and methods
  • Scalable sensitivity from analytical to preparative
  • High surface area silica for high preparative loading capacity
  • Impurity: <5 ppm metals