Stampa…
Tubing cutter for polymer tubing, Upchurch Scientific®
Fornitore: UPCHURCH SCIENTIFIC
A-327
554-3024EA 55.9 EUR
554-3024
Tubing cutter for polymer tubing, Upchurch Scientific®
taglierini
Designed to cut polymer tubing ¹/₁₆" and ¹/₈" O.D.
Complete with guide holes to ensure precise cutting. The cutter is durable, reliable and easy to operate.
Informazioni per l'ordine: Complete unit supplied with five replacement blades.