Stampa…
Reversed phase chromatography columns, RSpak JJ-50
Fornitore: Resonac
F7008220
554-2926EA 1059 EUR
554-2926
Reversed phase chromatography columns, RSpak JJ-50
Colonne cromatografiche
RSpak JJ-50 columns are ideal for analysis of complex samples containing neutral and ionic substances (surfactants) by multimode (reversed-phase and anion exchange) analysis.
  • Packing material contains trace amounts of quaternary ammonium groups
  • Base material is polyvinyl alcohol