Stampa…
TLC education advanced kit, micro-set F2
Fornitore: MACHEREY-NAGEL
814300 814301 814302
552-0288EA 177 EUR
552-0288 MANA814301 MANA814302
TLC education advanced kit, micro-set F2
Materiale didattico Materiali didattici di chimica Attività di apprendimento - Cromatografia
Advanced kit for science education.
  • All chemicals included for the separation of:
  • Edible fats
  • Fats and cholesterol in blood
Together with the materials from TLC micro-Set M samples of butter, margarine, and edible oil as well as blood can be separated and visualised by molybdatophosphoric acid spray reagent.
Attenzione: * These products contain harmful substances which must be specially labelled as hazardous.