Stampa…
Siringhe per microvolumi 10 µl
Fornitore: Trajan Scientific and Medical
002000 002003 002030 002033 002035 002050 002080 002100 002105 002130 002133 002135 002180 002400 002450 002839 002005 002450CAL
549-0279EA 50.4 EUR
549-0279 549-0430 549-0280 548-0198 548-0199 549-0281 549-0283 549-0284 549-0434 549-0285 548-0779 548-0934 549-0287 549-0290 549-0291 549-0429 549-0431 SGEA002450CAL
Siringhe per microvolumi 10 µl
Siringhe Siringhe per cromatografia
These 10 µl metal plunger syringes with stainless steel needle are ideal for making microvolume injections of 1,5 to 8 µl. Each plunger is individually fitted for each microsyringe for perfect sealing.
  • Accuracy and reproducibility: ±1%
  • Borosilicate glass barrel OD: 6,5 mm and 8 mm for guided plunger
  • Scale length: 54,1 mm
Plunger protection incorporates a plunger protection guide and plunger button reinforcement to minimise the risk of bent plungers.
SuperfleX microsyringes use a super elastic alloy plunger that will not bend or kink.