Stampa…

HMDS (1,1,1,3,3,3-Esametildisilazano) ≥99.0% (by GC), LiChropur™ for GC derivatisation, Supelco®

Fornitore: Merck

Synonyms: Hexamethyldisilazane

52619-250ML 52619-50ML
52619-250MLEA 284 EUR
52619-250ML 52619-50ML
HMDS (1,1,1,3,3,3-Esametildisilazano) ≥99.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
HMDS (1,1,1,3,3,3-Esametildisilazano)
Formula: (CH₃)₃SiNHSi(CH₃)₃
Punto di fusione: –82 °C
MDL Number: MFCD00008259
CAS Number: 999-97-3
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bb897a49260bc7677ad1f6d36b4d92877583818&vid=aacae6d820fe7f24349e379841e4cf581b4c637f&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR