Stampa…
Buffer, PBS tablets, pH 7,4, Millipore®
Fornitore: Merck Millipore (Oncogene)
524650-1
524650-1EA 90.6 EUR
524650-1
Buffer, PBS tablets, pH 7,4, Millipore®
Soluzioni tampone Tamponi per elettroforesi Tamponi di lavaggio
Developed for use in immunoassay applications.
Dissolving one tablet of PBS buffer in 1 litre of deionised water yields 140 mM NaCl, 10 mM phosphate buffer, and 3 mM KCl, pH 7,4 at 25 °C.