Stampa…
Analytical ultracentrifuge (AUC), Optima
Fornitore: Beckman Coulter
B86437 C00707 C00709 C00710
521-4196EA 0 EUR
521-4196 521-4203 521-4204 521-4205
Analytical ultracentrifuge (AUC), Optima
Centrifughe Centrifughe verticali a pavimento
The Optima AUC includes new features to improve the user experience from method setup to data recovery. Quick-turn absorbance and interference optics, paired with remote instrument setup and viewing, offer streamlined method setup and live data analysis. Importantly, the Optima AUC maintains compatibility with existing Beckman AUC rotors and accessories. Resolving similar components, such as viral particle capsids, can be challenging. However, the faster scan rate and improved radial resolution in the Optima AUC enable enhanced discrimination of such materials. Along with greater wavelengthMore Product Information