Stampa…
Microcentrifuga, universale ad alta velocità, 5430 / 5430 R (IVD)
Fornitore: EPPENDORF
5427000.011 5427000.216 5427000.062 5427000.267 5428000.015 5428000.210 5428000.414 5428000.619 5428000.066 5428000.260 5428000.465 5428000.660 5427000.410 5427000.615 5428000.716
521-2006EA 3891 EUR
521-2006 521-2007 521-2052 521-1280 521-1172 521-1173 521-1174 521-1175 521-1281 521-1282 521-1283 521-1284 521-1548 521-1645 521-1886
Microcentrifuga, universale ad alta velocità, 5430 / 5430 R (IVD)
Centrifughe Microcentrifughe
These unique cross-over centrifuges combine the best features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument.
  • Can be fitted with 12 different rotors for many applications (microtubes, PCR strips, cryotubes, conical tubes, PCR plates, deep-well plates, slides)
  • Five program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Soft-touch one-finger lid closure for ergonomic operation
  • Eppendorf QuickLock® – System for quickly opening and closing the rotor lid
  • Easy-to-use operator