Stampa…
Plastic coated filtration flasks, with assembly set made from PP
Fornitore: witeg Labortechnik
5.071.010 5.071.025 5.071.050 5.071.100 5.071.200 5.071.300 5.071.500 5.071.900 5.071.915 5.071.920
519-4121EA 39.4 EUR
519-4121 519-4122 519-4123 519-4124 519-4125 519-4221 519-4222 519-4223 519-4224 519-4225
Plastic coated filtration flasks, with assembly set made from PP
Fiasche Matracci filtranti
DURAN® with plastic sleeve, with hose connector assembly made from PP.
  • Splinter protection sleeve in cases of implosion
  • The sleeve remains transparent, firmly attached and durable up to 60 °C
  • Up to 2000 ml in Erlenmeyer form, from 3000 ml in bottle form
Vacuum-tight.
Certificazioni: DIN 12476 and ISO 6556