Stampa…
Capsule filters, Polycap™ TF, Whatman™
Fornitore: Whatman products (Cytiva)
6700-3602 6700-3610 6700-7501 6700-7502 6700-7504 6701-7510 6710-3602 6711-3602 6711-3604 6711-7502 2601T 2700T 2702T 2702M 2602S 2603 2603T 2703T 2800T 2801 2802T 6710-7502 2804T 2803T
515-0091EA 139 EUR
515-0091 515-0092 515-0093 515-0094 515-0095 515-0096 515-0152 516-4450 516-4451 516-4453 516-0911 516-0962 516-0913 516-0963 516-0955 516-0442 516-0956 516-0964 516-0973 516-0974 516-0975 516-0917 516-0977 516-0976
Capsule filters, Polycap™ TF, Whatman™
Filtri Capsule per filtrazione a membrana
These Polycap™ TF capsule filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a PP housing for use with organic solvents and chemically aggressive solutions.
  • PTFE filter membrane is resistant to most solvents
  • 1,0 µm pore size capsules used for extended life and filtration of highly contaminated solutions
  • Autoclavable capsule filtration device if sterility is required
Filtration of organic solvents and aggressive solutions requires a durable membrane for filtration without sample contamination.
Autoclavable Polycap™More Product Information