Stampa…
VWR®, Filtri in fibra di vetro
Fornitore: VWR Collection
516-0858EA 21.7 EUR
516-0858 516-0859 516-0860 516-0861 516-0862 516-0863 516-0864 516-0865 516-0866 516-0867 516-0868 516-0869 516-0870 516-0871 516-0872 516-0873 516-0874 516-0875 516-0876 516-0877 516-0879 516-0878 516-0881 516-0882 516-0883 516-0884 516-0885 516-0886 516-0887 516-0888 516-0890 516-0889 516-0343 516-0345 516-0344 516-0346 516-0347 516-0348
VWR®, Filtri in fibra di vetro
Filtri Filtri in fibra di vetro
Binder-free glass fibre filters feature fast flow rates, high loading capacity, wide thermal tolerance and excellent precipitate retention.
Grade 691: Ideal for radioimmunoassy of weak beta emitters by scintillation counting and for filtration of precipitated proteins, cells and cell debris. Also used for gravimetric determination of airborne particulates and air pollution monitoring.
Grade 692: Recommended for use in filtration of suspended solids in industrial effluents and also as pre-filters for membranes.
Grado 693: Utilizzato per la chiarificazione e il monitoraggio di solidiMore Product Information