Stampa…
Dispositivi per centrifugazione, Microsep™ Advance
Fornitore: Cytiva (Formerly Pall Lab)
MCP003C46 MCP003C41 MCP010C46 MCP030C41 MCP010C41 MCPM45C68 MCPM45C67 MCPM02C68 MCPM02C67 MCP100C46 MCP100C41 MCP030C46 MCP001C41 MCP001C46
516-0371EA 823 EUR
516-0371 516-0370 516-0373 516-0374 516-0372 516-0383 516-0382 516-0381 516-0378 516-0377 516-0376 516-0375 516-9920 516-0485
Dispositivi per centrifugazione, Microsep™ Advance
Filtri Filtri centrifughi
These centrifugal devices provide a precise, quick recovery of microlitre volumes of concentrate.
  • High recoveries, typically greater than 90%
  • Deadstop to prevent samples from spinning to dryness
  • Fits centrifuges that accept standard 17×100 mm tubes
Applications: Concentrate dilute protein samples prior to electrophoresis. Exchange buffer and remove salt in samples. Remove proteins and particulate from samples for HPLC analysis of drugs, amino acids, and antibodies. Isolate low molecular weight compounds from fermentation broths for natural productMore Product Information