Stampa…
VWR® Low Nitrogen Content Filters, Grade 521
Fornitore: VWR Collection
516-0316EA 24.7 EUR
516-0316
VWR® Low Nitrogen Content Filters, Grade 521
Filtri Carta da filtro Carta da filtro per analisi quantitative
Low nitrogen content filters for determination of nitrogen content in steel and iron samples dissolved in acid. Filtration of fine precipitates to dissolve according Kjeldahl method.