Stampa…
Grade 520 bII filter papers for technical use, Whatman™
Fornitore: Whatman products (Cytiva)
10331687
515-8032EA 1500 EUR
515-8032
Grade 520 bII filter papers for technical use, Whatman™
Filtri Carta da filtro Carta da filtro per analisi qualitative
A thick paper with high wet strength offering a very high flow rate.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.
Also available prepleated as Grade 520 bII ½.