Stampa…
Grade 602 h ½ qualitative filter papers, Whatman™
Fornitore: Whatman products (Cytiva)
10312642 10312644 10312645 10312647 10312651
515-5802EA 48.4 EUR
515-5802 515-5804 515-5805 515-5806 515-5807
Grade 602 h ½ qualitative filter papers, Whatman™
Filtri Carta da filtro Carta da filtro per analisi qualitative
A dense prepleated filter paper for collecting very small particles and removing fine precipitates.
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of