Stampa…
VWR®, Bottle-Top Vacuum Filtration Systems, PVDF, SFCA, Nylon
Fornitore: VWR Collection
514-1047EA 247 EUR
514-1047 514-1048 514-1012 514-1013 514-1014 514-1015 514-1016 514-1017 514-1018 514-1019 514-1020 514-1021 514-1022 514-1023 514-1024 514-1025 514-1026 514-1027 514-1028 514-1029 514-1030 514-1031 514-1032 514-1033 514-1034 514-1035 514-1036 514-1037 514-1038 514-1039 514-1040 514-1041 514-1042 514-1043 514-1044 514-1045 514-1046 514-1049 514-1050 514-1051 514-1052 514-1053 514-1054 514-1055 514-1056 514-1057 514-1058 514-1059 514-1060 514-1061 514-1062 514-1063 514-1064 514-1065 514-1066 514-1067
VWR®, Bottle-Top Vacuum Filtration Systems, PVDF, SFCA, Nylon
Filtri Filtri per bottiglie
These vacuum filter units are very useful in large volume samples separation and purification.
  • Large knurls on the reservoir bottle cap
  • Wide and easy access bottle mouth for efficient and stable pouring
  • Engraved graduation
  • Ergonomically designed side walls and collar
  • Hose connector can fit multiple hose diameters
  • Electron beam sterilised
  • Non pyrogenic
PVDF membrane: Extremely low protein binding; for filtration of non aggressive aqueous and mild organic solutions, or where maximising protein recovery is important.
Nylon membrane: Provides a broadMore Product Information
Visualizza i codici
Specifiche
Environmentally Preferable Produzione a basso impatto
Certificazione ambientale ISO 14001/Certificate No.: 04923E00243R0M