Stampa…
Filtri senza siringa, Autovial™, Whatman™
Fornitore: Whatman products (Cytiva)
AV125EAQU AV125ENAO AV125EORG AV125UAQU AV125UGMF AV125UNAO AV125UORG AV115NPEORG AV115NPUAQU AV115NPUORG AV115UGMF AV125NPUAQU AV525UAQU AV525UORG AV525UNAO
514-0013EA 374 EUR
514-0013 516-0452 514-0014 514-0015 514-0016 516-0460 514-0017 514-0326 516-0448 514-0327 516-0451 516-0454 516-0463 516-0465 516-0464
Filtri senza siringa, Autovial™, Whatman™
Filtri Filtri syringless
The Whatman Autovial Syringeless Filter replaces multiple syringe-coupled sample filtration devices with a single, disposable unit.
  • Available in 5 or 12 ml capacity
  • Single unit convenience – pre-assembled and easy to load
  • Self-contained, preventing filter pop-off with hazardous samples
  • Range of sterile or nonsterile membrane media available
  • Built-in air purge maximizes sample recovery
  • Glass fiber or polypropylene prefilter in selected 12 ml models – for difficult-to-filter samples
  • Choice of glass fiber or polypropylene prefilters
Autovial from Cytiva'More Product Information