Stampa…
Filtri in fibra di vetro
Fornitore: Cytiva (Formerly Pall Lab)
60010 61630 61631 61632 61633 61635 61638 61652 61655 61663 61664 64798 66075 66078 66085 66088 66198 66209 66211 66213 66215 66217 66224 66226 66227 66559 60127
516-7725EA 109 EUR
516-7725 513-0001 513-0002 516-7789 516-7790 513-0003 516-7792 513-0004 516-7795 516-7796 516-7797 513-0005 513-0007 513-0008 513-0009 516-7834 513-0011 513-0013 516-7863 513-0015 513-0017 516-7868 513-0022 513-0023 516-9016 516-7939 516-9108
Filtri in fibra di vetro
Filtri Filtri in fibra di vetro
Superior grade glass fibre filters for a variety of air sampling methods. Used in a variety of sample clean-up, prefiltration and analytical testing applications.
Type A/B: Binder-free borosilicate glass microfibres. For diagnostic applications and sample prefiltration. High loading capacity with 2,5 times thicker glass than type A/C.
Type A/C: Binder-free borosilicate glass microfibres. For testing dissolved and suspended solids in wastewater, for cell harvesting applications and prefiltration.
Grado A/D: Microfibre di vetro borosilicato senza materiale legante. PrefiltriMore Product Information