Stampa…
Filtri per siringa, Minisart® RC
Fornitore: Sartorius
17761-K 17761-Q 17762-K 17762-Q 17764-K 17764-Q 17764-S 17765-K 17765-Q 17765-S 17821-K 17821-Q 17822-K 17822-Q 17764-ACK 17761-ACK 17765-R 17761-R
512-3176EA 173 EUR
512-3176 512-3276 512-3178 512-3278 512-3180 512-3280 516-0908 512-3182 512-3282 514-5001 611-0716 512-3272 512-3174 510-3274 516-0907 516-0906 SART17765-R 516-0954
Filtri per siringa, Minisart® RC
Filtri Filtri per siringhe
Minisart® RC with hydrophilic regenerated cellulose (RC) is optimised for aqueous liquids and solvents with a pH of 3 - 14 and is stable against DMSO. With a pore size of 0,2 µm that can be used for sample preparation and sterile filtration.
  • Sterile and ready-to-use
  • Minimum extractables or leachables
  • Very low non-specific binding
  • Outperforming chemical compatibility
Individually packed and sterile Minisart® RC syringe filters are optimal for immediate usage whenever sterile filtration or clarification of aqueous and solvent samples is needed.