Stampa…
Grade 5 qualitative filter papers, Whatman™
Fornitore: Whatman products (Cytiva)
1005-042 1005-055 1005-070 1005-090 1005-110 1005-125 1005-150 1005-185 1005-240 1005-047 1005-320 1005-325
512-0476EA 14.2 EUR
512-0476 516-3505 512-1031 516-3508 512-1032 512-1033 512-1034 512-1035 516-0602 512-0475 512-0477 512-0478
Grade 5 qualitative filter papers, Whatman™
Filtri Carta da filtro Carta da filtro per analisi qualitative
A standard grade clarifying filter excellent for cloudy suspensions and for water and soil analysis.
  • Thickness of 2,5 µm
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
The maximum degree of fine particle filtration in the qualitative range. Capable of retaining the fine precipitates encountered in chemical analysis. Slow flow rate. ExcellentMore Product Information