Stampa…
MF-Millipore™ Membrane Filters
Fornitore: Merck
AAWP02500 AAWP03700 AAWP04700 GSWP02500 GSWP04700 HATF04700 HATF08250 HAWP03700 HAWP04700 PHWP04700 RAWG02500 RAWP01300 SMWP01300 SMWP04700 SSWP14250 VCWP04700 RAWP04700 AABP04700 AABG01300 AABG02500 AABG03700 AABG04700 AABP02500 AAWG01300 AAWG02500 AAWG0250C AAWG03700 AAWG04700 AAWP01300 AAWP037P0 AAWP037PM AAWP0470M AAWP09000 AAWP14250 DAWP01300 DAWP02500 DAWP09025 DAWP14250 GSTF01300 GSTF02500 GSTF09000 GSTF14250 GSWP03700 GSWP09000 HABG01300 HABG02500 HABG04700 HABP02500 HABP04700 HATF01300 HATF02500 HATF09025 HATF14250 HAWG01300 HAWG02500 HAWG03700 HAWG04700 HAWP02400 HAWP0470M PHWP02500 PHWP09025 PHWP14250 HAWP09000 RATF04700 RATF14250 RAWG0250C RAWG04700 RAWP03700 SSWP01300 VSWP14250 VSWP04700 SCWP01300 SCWP02500 SCWP09025 SCWP14250 SMWP0190R SMWP02500 SMWP03700 SMWP09025 SMWP14250 SSWP02500 SSWP09025 VCWP01300 VCWP02500 VCWP05000 VCWP09025 VCWP14250 VMWP01300 VMWP02500 VMWP04700 VMWP09025 VSWP09025 SSWP04700 SCWP04700 GSWP01300 VSWP02500 HAWP14250 GSTF04700 GSWP14250 RAWP02500 DAWP04700 RAWP14250 HAWP01300 VSWP01300 HAWP05000 HATF08550 HAWP02500 RAWP05000 RAWP09025 SCWP0190R HAEP04700 HAWG04705 AABPC5000 HAWP04700F1
512-0240EA 150 EUR
512-0240 MILFAAWP03700 512-0241 512-0250 512-0251 MILFHATF04700 MILFHATF08250 MILFHAWP03700 512-0258 MILFPHWP04700 MILFRAWG02500 MILFRAWP01300 512-0273 512-0275 MILFSSWP14250 MILFVCWP04700 512-0267 MILFAABP04700 MILFAABG01300 MILFAABG02500 MILFAABG03700 MILFAABG04700 MILFAABP02500 MILFAAWG01300 MILFAAWG02500 MILFAAWG0250C MILFAAWG03700 MILFAAWG04700 MILFAAWP01300 MILFAAWP037P0 MILFAAWP037PM MILFAAWP0470M MILFAAWP09000 MILFAAWP14250 MILFDAWP01300 MILFDAWP02500 MILFDAWP09025 MILFDAWP14250 MILFGSTF01300 MILFGSTF02500 MILFGSTF09000 MILFGSTF14250 MILFGSWP03700 MILFGSWP09000 MILFHABG01300 MILFHABG02500 MILFHABG04700 MILFHABP02500 MILFHABP04700 MILFHATF01300 MILFHATF02500 MILFHATF09025 MILFHATF10050 MILFHATF14250 MILFHAWG01300 MILFHAWG02500 MILFHAWG03700 MILFHAWG04700 MILFHAWG05000 MILFHAWP02400 MILFHAWP0470M MILFPHWP02500 MILFPHWP09025 MILFPHWP14250 MILFHAWP09000 MILFRATF04700 MILFRATF14250 MILFRAWG0250C MILFRAWG04700 MILFRAWP03700 MILFSSWP01300 MILFVSWP14250 512-0283 MILFSCWP01300 512-0270 MILFSCWP09025 MILFSCWP14250 MILFSMWP0190R 512-0274 MILFSMWP03700 MILFSMWP09025 MILFSMWP14250 512-0276 MILFSSWP09025 MILFVCWP01300 MILFVCWP02500 MILFVCWP05000 MILFVCWP09025 MILFVCWP14250 MILFVMWP01300 MILFVMWP02500 512-0281 MILFVMWP09025 MILFVSWP09025 512-0277 512-0271 512-0249 512-0282 MILFHAWP14250 MILFGSTF04700 MILFGSWP14250 MILFRAWP02500 MILFDAWP04700 MILFRAWP14250 512-0256 MILFVSWP01300 MILFHAWP05000 MILFHATF08550 512-0257 MILFRAWP05000 MILFRAWP09025 MILFSCWP0190R MILFHAEP04700 MILFHAWG04705 MILFAABPC5000 MILFHAWP04700F1
MF-Millipore™ Membrane Filters
Filtri Membrane filtranti
MF-Millipore™ filters without Triton® surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Biologically inert mixtures of cellulose acetate and cellulose nitrate.