Stampa…
Microbiological filtration system, MBS I, Whatman™
Fornitore: Whatman products (Cytiva)
10477602 10445868 10445863 10445871
515-0177EA 52.9 EUR
515-0177 511-0143 511-0140 WHAT10445871
Microbiological filtration system, MBS I, Whatman™
Filtri Imbuti filtranti a membrana
MBS I is an excellent system for optimal microbiological control using membranes. The overall procedure time is reduced to a minimum. The design of the system, which consists of an electrical membrane dispenser, a funnel dispenser, and a vacuum manifold, leads to more reproducible results. All components are sold separately. The modular system provides flexibility to select the hardware and accessories that best suit the unique and varied workflows of the user.
The special sealing technique ensures easy handling and high integrity of the funnel and membrane during filtration. This keeps anyMore Product Information