Stampa…
Test sieves, 203×50 mm
Fornitore: Retsch GmbH
60.149.000020 60.149.000025 60.149.000032 60.149.000036 60.149.000038 60.149.000040 60.149.000045 60.149.000050 60.149.000053 60.149.000056 60.149.000063 60.149.000071 60.149.000075 60.149.000080 60.149.000090 60.149.000100 60.149.000106 60.149.000112 60.149.000125 60.149.000140 60.149.000150 60.149.000160 60.149.000180 60.149.000200 60.149.000212 60.149.000224 60.149.000250 60.149.000280 60.149.000300 60.149.000315 60.149.000355 60.149.000400 60.149.000425 60.149.000450 60.149.000500 60.149.000560 60.149.000600 60.149.000630 60.149.000710 60.149.000800 60.149.000850 60.149.000900 60.149.001000 60.149.001120 60.149.001180 60.149.001250 60.149.001400 60.149.001600 60.149.001700 60.149.001800 60.149.002240 60.149.002360 60.149.002500 60.149.002800 60.149.003150 60.149.003350 60.149.003550 60.149.004000 60.149.004500 60.149.004750 60.149.005000 60.149.005600 60.149.006300 60.149.006700 60.149.007100 60.149.008000 60.149.009000 60.149.009500 60.149.010000 60.149.011200 60.149.012500 60.149.013200 60.149.014000 60.149.016000 60.149.018000 60.149.019000 60.149.020000 60.149.022400 60.149.025000 60.149.026500 60.149.028000 60.149.031500 60.149.035500 60.149.037500 60.149.040000 60.149.045000 60.149.050000 60.149.053000 60.149.056000 60.149.063000 60.149.071000 60.149.075000 60.149.080000 60.149.090000 60.149.100000 60.149.106000 60.149.112000 60.149.125000 60.149.002000
510-4701EA 914 EUR
510-4701 510-4702 510-4703 510-4704 510-4705 510-4706 510-4707 510-4708 510-4709 510-4710 510-4711 510-4712 510-4713 510-4714 510-4715 510-4716 510-4717 510-4718 510-4719 510-4720 510-4721 510-4722 510-4723 510-4724 510-4725 510-4726 510-4727 510-4728 510-4729 510-4730 510-4731 510-4732 510-4733 510-4734 510-4735 510-4736 510-4737 510-4738 510-4739 510-4740 510-4741 510-4742 510-4743 510-4744 510-4745 510-4746 510-4747 510-4748 510-4749 510-4750 510-4752 510-4753 510-4754 510-4755 510-4756 510-4757 510-4758 510-4759 510-4760 510-4761 510-4762 510-4763 510-4764 510-4765 510-4766 510-4767 510-4768 510-4769 510-4770 510-4771 510-4772 510-4773 510-4774 510-4775 510-4776 510-4777 510-4778 510-4779 510-4780 510-4781 510-4782 510-4783 510-4784 510-4785 510-4786 510-4787 510-4788 510-4789 510-4790 510-4791 510-4792 510-4793 510-4794 510-4795 510-4796 510-4797 510-4798 510-4799 510-4751
Test sieves, 203×50 mm
Setacci Setacci per analisi
304 stainless steel, ISO 3310-1:2001-09.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieves are individually laser engraved with clear and precise labelling for full traceability. Available in three of the most commonly used sizes. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.