Stampa…
VWR®, Test Sieves, 76×22 mm
Fornitore: VWR Collection
510-3611EA 897 EUR
510-3611 510-3612 510-3613 510-3614 510-3615 510-3616 510-3617 510-3618 510-3619 510-3620 510-3621 510-3622 510-3623 510-3624 510-3625 510-3626 510-3627 510-3628 510-3629 510-3630 510-3631 510-3632 510-3633 510-3634 510-3635 510-3636 510-3637 510-3638 510-3639 510-3640 510-3641 510-3642 510-3643 510-3644 510-3645 510-3646 510-3647 510-3648 510-3649 510-3650 510-3651 510-3652 510-3653 510-3654 510-3655 510-3656 510-3657 510-3658 510-3659 510-3660 510-3661 510-3814 510-3815 510-3816
VWR®, Test Sieves, 76×22 mm
Setacci Setacci per analisi
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informazioni perMore Product Information