Stampa…
VWR®, Test Sieves, 400×65 mm, with Square Holes
Fornitore: VWR Collection
510-3570EA 390 EUR
510-3570 510-3571 510-3572 510-3573 510-3574 510-3575 510-3576 510-3577 510-3578 510-3579 510-3580 510-3581 510-3582 510-3583 510-3584 510-3585 510-3586 510-3587 510-3588 510-3589 510-3590 510-3591 510-3592 510-3593 510-3594 510-3595 510-3596 510-3597 510-3598 510-3599 510-3600 510-3601 510-3602 510-3603 510-3604 510-3605 510-3606 510-3607 510-3608 510-3609
VWR®, Test Sieves, 400×65 mm, with Square Holes
Setacci Setacci per analisi
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
InformazioniMore Product Information