Stampa…
VWR®, Test Sieves, 305×60 mm
Fornitore: VWR Collection
510-2837EA 266 EUR
510-2837 510-2838 510-2839 510-2840 510-2841 510-2842 510-2843 510-2844 510-2845 510-2846 510-2847 510-2848 510-2849 510-2850 510-2851 510-2852 510-2853 510-2854 510-2855 510-2856 510-2857 510-2858 510-2859 510-2860 510-2861 510-2862 510-2863 510-2864 510-2865 510-2866 510-2867 510-2868 510-2869 510-2870 510-2871 510-2872 510-2873 510-2874 510-2875 510-2876 510-2877 510-2878 510-2879 510-2880 510-2881 510-2882 510-2883 510-2884 510-2885 510-2886 510-2887 510-2888 510-2889 510-2890 510-2891 510-2892 510-2893 510-2894 510-2895 510-2896 510-2897 510-2898 510-2899 510-2900 510-2901 510-2902 510-2903 510-2904 510-2905 510-2906 510-2907 510-2908 510-2909 510-2910 510-2911 510-2912 510-2913 510-2914 510-2915 510-2916 510-2917 510-2918 510-2919 510-2920 510-2921 510-2922 510-2923 510-2924 510-2925 510-2926 510-2927 510-2928
VWR®, Test Sieves, 305×60 mm
Setacci Setacci per analisi
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informazioni perMore Product Information