Stampa…
VWR®, Setacci per analisi, 203×32 mm
Fornitore: VWR Collection
510-0840EA 793 EUR
510-0840 510-0841 510-0842 510-0843 510-0844 510-0845 510-0846 510-0847 510-0848 510-0849 510-0850 510-0851 510-0852 510-0853 510-0854 510-0855 510-0856 510-0857 510-0858 510-0859 510-0860 510-0861 510-0862 510-0863 510-0864 510-0865 510-0866 510-0867 510-0868 510-0869 510-0870 510-0871 510-0872 510-0873 510-0874 510-0875 510-0876 510-0877 510-0878 510-0879 510-0880 510-0881 510-0882 510-0883 510-0884 510-0885 510-0886 510-0887 510-0888 510-0889 510-0890 510-0891 510-0892 510-0893 510-0894 510-0895 510-0896 510-0897 510-0898 510-0899 510-0900 510-0901 510-0902 510-0903 510-0904 510-0905 510-0906 510-0907 510-0908 510-0909 510-0910
VWR®, Setacci per analisi, 203×32 mm
Setacci Setacci per analisi
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informazioni perMore Product Information