Stampa…
VWR®, Test Sieves, 100×45 mm
Fornitore: VWR Collection
510-0526EA 156 EUR
510-0526 510-0527 510-0528 510-0529 510-0530 510-0531 510-0532 510-0533 510-0534 510-0535 510-0536 510-0537 510-0500 510-0501 510-0502 510-0503 510-0504 510-0505 510-0506 510-0507 510-0508 510-0509 510-0510 510-0511 510-0512 510-0513 510-0514 510-0515 510-0516 510-0517 510-0518 510-0519 510-0520 510-0521 510-0522 510-0523 510-0524 510-0525 510-0538 510-0539 510-0540 510-0541 510-0542 510-0543 510-0544 510-0545 510-0546 510-0547 510-0548 510-0549 510-0550 510-0551 510-0552 510-0553
VWR®, Test Sieves, 100×45 mm
Setacci Setacci per analisi
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informazioni perMore Product Information