Stampa…
Avantor® Apex II Amino, HPLC Columns
Fornitore: Avantor
4M15344EA 854 EUR
4M15344
Avantor® Apex II Amino, HPLC Columns
Colonne cromatografiche
Avantor® Apex HPLC column phases are recommended for routine analyses.
  • Conventional 100 Å pore size spherical silica
  • Narrow particle distribution
  • Controlled surface area
  • USP L8