Stampa…

Anti-Nitric Oxide Synthase (1131-1144), Inducible Rabbit Polyclonal Antibody

Fornitore: Merck Millipore (Oncogene)
482728-100
482728-100EA 925 EUR
482728-100
Anti-Nitric Oxide Synthase (1131-1144), Inducible Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Nitric Oxide Synthase (1131-1144), Inducible
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR